Please login to your online account to display your discounted pricing

Mercuric Chloride, 5% (w/v) Aqueous, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR465000020F

 View more versions of this product

 Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Catalog No. R465000020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 20L
Chemical Name or Material Mercuric Chloride
CAS 7732-18-5
CAS 7487-94-7
CAS Min % 93.0
Packaging Cubitainer
CAS Max % 97.0
CAS Max % 4.86
CAS Min % 4.67

Specifications

Chemical Name or Material Mercuric Chloride
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7487-94-7
CAS Min % 4.67
CAS Max % 4.86
Physical Form Liquid