Please login to your online account to display your discounted pricing

β-Mercaptoethylamine Hydrochloride, Fisher BioReagents


Quantity 25g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP266425

 View more versions of this product

Catalog No. BP266425

  • / Each
  • / Case of 8 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 113.61
Melting Point 66°C
Quantity 25g
Merck Index 15, 2776
Physical Form Solid
Molecular Formula C2H7NS · HCl
Color White
pH 3.3 to 5.0
Synonym 2-Mercaptoethylamine HCl
Assay Percent Range 98%
Packaging Amber Glass
Purity Approx. 98%
Health Hazard 3 Emergency Overview
Harmful if swallowed. Hygroscopic. Stench. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:1
Instability:1
Health Hazard 2 WARNING!
ChemAlert Storage Symbol Gray
Recommended Storage 4°C, desiccate
CAS 156-57-0
Chemical Name or Material β-Mercaptoethylamine Hydrochloride

Specifications

Chemical Name or Material β-Mercaptoethylamine Hydrochloride
CAS 156-57-0
Physical Form Solid
Assay Percent Range 98%
Molecular Formula C2H7NS · HCl
Formula Weight 113.61
Synonym 2-Mercaptoethylamine HCl
Accessories
 

Emergency Overview
Harmful if swallowed. Hygroscopic. Stench. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:1
Instability:1

WARNING!

4°C, desiccate

SDS & Safety Documents