missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

β-Mercaptoethylamine Hydrochloride, Fisher BioReagents

$132.02 - $792.78

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym 2-aminoethanethiol hydrochloride, 2-mercaptoethylamine hcl, 2-mercaptoethylamine hydrochloride, bekaptan, cysteamine hcl, cysteamine hydrochloride, cysteaminium chloride, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
IUPAC Name 2-aminoethanethiol;hydrochloride
SMILES [H+].[Cl-].NCCS
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP26645
SDS View Documents Promotion Details
Fisher BioReagents
BP26645
5g Amber Glass
Each for $132.02
Case of 9 Each for $1,188.18
 
BP266425
SDS View Documents Promotion Details
Fisher BioReagents
BP266425
25g Amber Glass
Each for $199.27
Case of 8 Each for $1,594.16
 
BP2664100
SDS View Documents Promotion Details
Fisher BioReagents
BP2664100
100g Amber Glass
Each for $792.78
Case of 9 Each for $7,135.02
 
Description

Description

β-Mercaptoethylamine HCl is used as an antioxidant in several biochemical procedures.
Specifications

Chemical Identifiers

156-57-0
113.60
OGMADIBCHLQMIP-UHFFFAOYSA-N
9082
[H+].[Cl-].NCCS
C2H8ClNS
MFCD00012904
2-aminoethanethiol hydrochloride, 2-mercaptoethylamine hcl, 2-mercaptoethylamine hydrochloride, bekaptan, cysteamine hcl, cysteamine hydrochloride, cysteaminium chloride, mercaptamine hydrochloride, merkamin hydrochloride
2-aminoethanethiol;hydrochloride

Specifications

Amber Glass
66°C
3.3 to 5.0
98%
MFCD00012904
2-aminoethanethiol hydrochloride, 2-mercaptoethylamine hcl, 2-mercaptoethylamine hydrochloride, bekaptan, cysteamine hcl, cysteamine hydrochloride, cysteaminium chloride, mercaptamine hydrochloride, merkamin hydrochloride
[H+].[Cl-].NCCS
113.60
113.61
∽98%
White
5g
156-57-0
C2H8ClNS
15, 2776
OGMADIBCHLQMIP-UHFFFAOYSA-N
2-aminoethanethiol;hydrochloride
9082
Solid
β-Mercaptoethylamine Hydrochloride
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. Hygroscopic. Stench. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:1
Instability:1

Recommended Storage : 4°C, desiccate

SDS
Product Certifications