missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

β-Mercaptoethylamine Hydrochloride, Fisher BioReagents

Manufacturer:  Fisher BioReagents BP2664100

 View more versions of this product

Catalog No. BP2664100




Description

Description

β-Mercaptoethylamine HCl is used as an antioxidant in several biochemical procedures.
Specifications

Specifications

β-Mercaptoethylamine Hydrochloride
White
100g
156-57-0
C2H8ClNS
2-aminoethanethiol hydrochloride, 2-mercaptoethylamine hcl, 2-mercaptoethylamine hydrochloride, bekaptan, cysteamine hcl, cysteamine hydrochloride, cysteaminium chloride, mercaptamine hydrochloride, merkamin hydrochloride
C(CS)N.Cl
113.603
113.61
∽98%
Amber Glass
66°C
3.3 to 5.0
98%
15, 2776
OGMADIBCHLQMIP-UHFFFAOYSA-N
2-aminoethanethiol;hydrochloride
9082
Solid

Chemical Identifiers

156-57-0
113.603
2-aminoethanethiol hydrochloride, 2-mercaptoethylamine hcl, 2-mercaptoethylamine hydrochloride, bekaptan, cysteamine hcl, cysteamine hydrochloride, cysteaminium chloride, mercaptamine hydrochloride, merkamin hydrochloride
2-aminoethanethiol;hydrochloride
C2H8ClNS
OGMADIBCHLQMIP-UHFFFAOYSA-N
9082
C(CS)N.Cl
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. Hygroscopic. Stench. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:1
Instability:1

Recommended Storage : 4°C, desiccate

SDS Documents
Promotions

Promotions