missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercaptoacetic acid, sodium salt, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC148892500

 View more versions of this product

Catalog No. AC148892500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Mercaptoacetic acid, sodium salt
>300.0°C
98%
98%
HSCH2CO2Na
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium;2-sulfanylacetate
23690444
114.09
Glass bottle
250g
367-51-1
C2H3NaO2S
MFCD00043386
Solubility in water: 1000g/L (20°C). Other solubilities: slightly soluble in alcohol
C(C(=O)[O-])S.[Na+]
114.094
CHEBI:86481
98%

Chemical Identifiers

367-51-1
114.094
GNBVPFITFYNRCN-UHFFFAOYSA-M
23690444
sodium;2-sulfanylacetate
C2H3NaO2S
MFCD00043386
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
CHEBI:86481
C(C(=O)[O-])S.[Na+]
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Toxic in contact with skin.
May cause an allergic skin reaction.
May be corrosive to metals.

GHS P Statement
Keep only in original container.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Absorb spillage to prevent material damage.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 206-696-4

RTECSNumber : AI7700000

TSCA : TSCA

SDS Documents