Learn More
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 148890500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mercaptoacetic acid, sodium salt | |
367-51-1 | |
97.5% min. (Iodimetry) | |
C2H3NaO2S | |
MFCD00043386 | |
15,8819 | |
Solubility in water: 1000g/L (20°C). Other solubilities: slightly soluble in alcohol | |
C(C(=O)[O-])S.[Na+] | |
114.09 | |
CHEBI:86481 | |
98% |
98% | |
>300.0°C | |
Glass bottle | |
HSCH2CO2Na | |
50 g | |
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
GNBVPFITFYNRCN-UHFFFAOYSA-M | |
sodium;2-sulfanylacetate | |
23690444 | |
114.09 |
Chemical Identifiers
367-51-1 | |
114.09 | |
GNBVPFITFYNRCN-UHFFFAOYSA-M | |
23690444 | |
sodium;2-sulfanylacetate |
C2H3NaO2S | |
MFCD00043386 | |
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
CHEBI:86481 | |
C(C(=O)[O-])S.[Na+] |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin
GHS Signal Word: Danger
EINECSNumber : 206-696-4
RTECSNumber : AI7700000
TSCA : TSCA
RUO – Research Use Only