Please login to your online account to display your discounted pricing

Alfa Aesar™ Mercaptoacetic acid calcium salt trihydrate, 98%

$39.78 - $150.07

Chemical Identifiers

Chemical Name or Material Mercaptoacetic acid calcium salt trihydrate
CAS 65208-41-5
Molecular Formula C4H6CaO4S2·3H2O
Linear Formula (HSCH2CO2)Ca·3H2O
Formula Weight 276.34 (222.29 Anhydrous)
MDL Number MFCD00150707
Beilstein 3755791
Merck Index 14,1711
Synonym Calcium mercaptoacetate trihydrate; Calcium thioglycolate trihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4555709

  SDS
alfa aesar™
4555709
10g Each for $39.78

AA4555718

  SDS
alfa aesar™
4555718
50g Each for $150.07
Description & Specifications

Specifications

Formula Weight 276.34 (222.29 Anhydrous)
Chemical Name or Material Mercaptoacetic acid calcium salt trihydrate
TSCA TSCA
Physical Form Powder
Assay 98%
Molecular Formula C4H6CaO4S2·3H2O
Linear Formula (HSCH2CO2)Ca·3H2O
MDL Number MFCD00150707
CAS 65208-41-5
Beilstein 3755791
Merck Index 14,1711
Synonym Calcium mercaptoacetate trihydrate; Calcium thioglycolate trihydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Melting Point 280° to 290°C (dec.)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.