Learn More
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125432500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Mercaptoacetic acid | |
-16.0°C | |
1.3250g/mL | |
126°C | |
97.5% min. (Iodimetry) | |
C2H4O2S | |
HSCH2CO2H | |
250 mL | |
1.325 | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
2-sulfanylacetic acid | |
1133 | |
CHEBI:30065 | |
98% |
68-11-1 | |
Colorless | |
220.0°C | |
Authentic | |
Glass bottle | |
1.5030 to 1.5050 | |
MFCD00004876 | |
01,1153; 07,366 | |
15, 9489 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chloroform and benzene | |
C(C(=O)O)S | |
92.11 | |
6.55 Pas (20°C) | |
92.11 | |
Liquid |
Chemical Identifiers
68-11-1 | |
92.11 | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
1133 | |
2-sulfanylacetic acid |
C2H4O2S | |
MFCD00004876 | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
CHEBI:30065 | |
C(C(=O)O)S |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre
GHS Signal Word: Danger
EINECSNumber : 200-677-4
RUO – Research Use Only