missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mer (MerTK) Rat anti-Mouse, PE, Clone: DS5MMER, eBioscience™

Rat Monoclonal Antibody

$145.00 - $413.25

Specifications

Antigen Mer (MerTK)
Clone DS5MMER
Host Species Rat
Gene Alias Mer
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122327 Life Technologies
LS12575180
25μg Each for $145.00
501122328 Life Technologies
LS12575182
100μg Each for $413.25
Specifications

Description

This DS5MMER monoclonal antibody recognizes mouse MerTK, a 170-210kDa member of the TAM family of tyrosine kinase receptors that also includes Axl and Tyro3. MerTK is expressed on tissue macrophages and is involved in the removal of apoptotic cells. This process relies on two soluble ligands of MerTK, Protein S and Gas6 that bind to phosphatidylserine found on the outer leaflet of the plasma membrane of cells undergoing apoptosis. Upon binding these ligands, MerTK undergoes autophosphorylation at multiple tyrosine residues that activate the PI3K and Akt pathways. This results in the phagocytosis of apoptotic cells and also results in the direct inhibition of TLR-induced production of pro-inflammatory cytokines. In addition MerTK may function as a putative entry receptor for filoviruses. Deficiency of MerTK causes general autoimmunity, inflammation and accumulation of apoptotic bodies. MerTK is constitutively released from the cell surface by metalloproteinases and thus is present in the serum and culture medium. This process can be enhanced by stimulation with LPS. MerTK can be used to help discriminate macrophages from dendritic cells. MerTK is often expressed on malignant cells and may be implicated in immune evasion.

The DS5MMER antibody will work on paraformaldehyde-fixed cells.
Description & Specifications

Specifications

Mer (MerTK)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
DS5MMER
Mer
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL