Please login to your online account to display your discounted pricing

Tocris™ Meptazinol hydrochloride

mu1 partial agonist

$102.98 - $447.38

Specifications

Product Type Meptazinol hydrochloride
Inhibitors Meptazinol hydrochloride
Molecular Weight 269.81
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

363610R

 
tocris™
363610
10mg Each for $102.98

363650R

 
tocris™
363650
50mg Each for $447.38
Description & Specifications

Specifications

Product Type Meptazinol hydrochloride
Inhibitors Meptazinol hydrochloride
Molecular Weight 269.81
Purity >99

Centrally active opioid analgesic. Partial agonist at theμ1 opioid receptor. Also shown to inhibit acetylcholinesterase.

  • Chemical Name: 3-(3-Ethylhexahydro-1-methyl-1H-azepin-3-yl)phenol hydrochloride
  • Formula: C15H23NO.HCl
  • CAS Number: 59263-76-2
  • Solubility: Soluble to 100mM in water and to 75mM in DMSO
  • Purity: >99