missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mepivacaine Hydrochloride, TCI America™

$180.28 - $629.45

Chemical Identifiers

CAS 1722-62-9
Molecular Formula C15H23ClN2O
Molecular Weight (g/mol) 282.812
MDL Number MFCD00144738
InChI Key RETIMRUQNCDCQB-UHFFFAOYSA-N
Synonym N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride
PubChem CID 66070
ChEBI CHEBI:6760
IUPAC Name N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
SMILES CC1=C(C(=CC=C1)C)NC(=O)C2CCCCN2C.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25601G View Documents TCI America
M25601G
1g Each for $180.28
 
M25605G View Documents TCI America
M25605G
5g Each for $629.45
 
Specifications

Chemical Identifiers

1722-62-9
282.812
RETIMRUQNCDCQB-UHFFFAOYSA-N
66070
N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
C15H23ClN2O
MFCD00144738
N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride
CHEBI:6760
CC1=C(C(=CC=C1)C)NC(=O)C2CCCCN2C.Cl

Specifications

White
1g
C15H23ClN2O
N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide Hydrochloride
CC1=C(C(=CC=C1)C)NC(=O)C2CCCCN2C.Cl
282.812
CHEBI:6760
Crystal-Powder at 20°C
Mepivacaine Hydrochloride
263°C
1722-62-9
MFCD00144738
RETIMRUQNCDCQB-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide;hydrochloride
66070
282.81
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : TK6475000

TSCA : Yes