missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Menthyl Chloroformate, TCI America™

$70.28 - $209.31

Chemical Identifiers

CAS 14602-86-9
Molecular Formula C11H19ClO2
Molecular Weight (g/mol) 218.721
MDL Number MFCD00009694
InChI Key KIUPCUCGVCGPPA-KXUCPTDWSA-N
Synonym 1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
PubChem CID 7014897
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] carbonochloridate
SMILES CC1CCC(C(C1)OC(=O)Cl)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M09905ML View Documents TCI America
M09905ML
5mL Each for $70.28
 
M099025ML View Documents TCI America
M099025ML
25mL Each for $209.31
 
Specifications

Chemical Identifiers

14602-86-9
218.721
KIUPCUCGVCGPPA-KXUCPTDWSA-N
7014897
CC1CCC(C(C1)OC(=O)Cl)C(C)C
C11H19ClO2
MFCD00009694
1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] carbonochloridate

Specifications

109°C
5mL
C11H19ClO2
3277
KIUPCUCGVCGPPA-KXUCPTDWSA-N
[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] carbonochloridate
7014897
Clear Liquid at 20°C
(-)-Menthyl Chloroformate
Yellow
14602-86-9
MFCD00009694
1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
CC1CCC(C(C1)OC(=O)Cl)C(C)C
218.721
218.72
≥97.0% (T)
Safety and Handling

Safety and Handling

TSCA : No