missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-Menthol 98.0+%, TCI America™

Manufacturer:  TCI America M032125ML

 View more versions of this product

Catalog No. M032125ML



Specifications

Specifications

(+/-)-Menthol
25mL
C10H20O
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
CC1CCC(C(C1)O)C(C)C
156.269
156.27
≥98.0% (GC)
White
89-78-1
MFCD00001484
NOOLISFMXDJSKH-IVZWLZJFSA-N
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
6566020
Crystal at 20°C

Chemical Identifiers

89-78-1
156.269
NOOLISFMXDJSKH-IVZWLZJFSA-N
6566020
CC1CCC(C(C1)O)C(C)C
C10H20O
MFCD00001484
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2333

RTECSNumber : OT0525000

TSCA : Yes

Documents