missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 15356-60-2 |
---|---|
Molecular Formula | C10H20O |
Molecular Weight (g/mol) | 156.269 |
MDL Number | MFCD00062983 |
InChI Key | NOOLISFMXDJSKH-AEJSXWLSSA-N |
Synonym | +-menthol, d-menthol, 1s,2r,5s-+-menthol, 1s,2r,5s-menthol, racementhol, 1s,2r,5s-2-isopropyl-5-methylcyclohexanol, unii-c6b1oe8p3w, dl-menthol, menthol, +, cyclohexanol, 5-methyl-2-1-methylethyl-, 1s,2r,5s |
PubChem CID | 165675 |
ChEBI | CHEBI:76306 |
IUPAC Name | (1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
SMILES | CC1CCC(C(C1)O)C(C)C |
Chemical Identifiers
15356-60-2 | |
156.269 | |
NOOLISFMXDJSKH-AEJSXWLSSA-N | |
165675 | |
(1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
C10H20O | |
MFCD00062983 | |
+-menthol, d-menthol, 1s,2r,5s-+-menthol, 1s,2r,5s-menthol, racementhol, 1s,2r,5s-2-isopropyl-5-methylcyclohexanol, unii-c6b1oe8p3w, dl-menthol, menthol, +, cyclohexanol, 5-methyl-2-1-methylethyl-, 1s,2r,5s | |
CHEBI:76306 | |
CC1CCC(C(C1)O)C(C)C |
Specifications
99°C | |
42°C | |
15356-60-2 | |
MFCD00062983 | |
NOOLISFMXDJSKH-AEJSXWLSSA-N | |
(1S,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol | |
165675 | |
156.27 | |
≥99.0% (GC) |
White | |
100g | |
C10H20O | |
+-menthol, d-menthol, 1s,2r,5s-+-menthol, 1s,2r,5s-menthol, racementhol, 1s,2r,5s-2-isopropyl-5-methylcyclohexanol, unii-c6b1oe8p3w, dl-menthol, menthol, +, cyclohexanol, 5-methyl-2-1-methylethyl-, 1s,2r,5s | |
CC1CCC(C(C1)O)C(C)C | |
156.269 | |
CHEBI:76306 | |
Crystal-Powder at 20°C | |
(+)-Menthol |
Safety and Handling
EINECSNumber : (3)-2333
RTECSNumber : OT0525000
TSCA : Yes