Please login to your online account to display your discounted pricing

MEM chloride, 94%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  185980100

 View more versions of this product

Catalog No. AC185980100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥92%
Physical Form Liquid
Formula Weight 124.57
Quantity 10g
Color Clear colorless to yellow
Fieser 07,227; 09,299
Density 1.09g/mL
CAS 542-88-1
Synonym β-Methoxyethoxymethylechloride
Molecular Formula C4H9ClO2
MDL Number MFCD00000888
Assay Percent Range 94%
CAS 3970-21-6
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause cancer.
Flammable liquid and vapor.
Refractive Index 1.4260 to 1.4280 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula CH3OCH2CH2OCH2Cl
Chemical Name or Material MEM chloride
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material MEM chloride
CAS 3970-21-6
CAS 542-88-1
Assay ≥92%
Assay Percent Range 94%
Molecular Formula C4H9ClO2
Linear Formula CH3OCH2CH2OCH2Cl
Formula Weight 124.57
MDL Number MFCD00000888
Synonym β-Methoxyethoxymethylechloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause cancer.
Flammable liquid and vapor.

SDS & Safety Documents