missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Melt and Cast Agarose Gels
Manufacturer: Fisher Scientific AZ950113
Description
Electrophoresis kit refills.
Specifications
0.8% | |
Agarose Gel Cassette | |
Agarose |
Liquid | |
DNA separation by gel electrophoresis | |
400 mL |