missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Melt and Cast Agarose Gels

$45.08 - $45.08

Specifications

Concentration 0.8%
For Use With (Application) DNA separation by gel electrophoresis
Material Agarose
Format Liquid
Product Type Agarose Gel Cassette
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S67095 View Documents Fisher Scientific
AZ950113
400 mL Each for $45.08
 
Description

Description

Electrophoresis kit refills.

Specifications

Specifications

0.8%
Agarose
Agarose Gel Cassette
DNA separation by gel electrophoresis
Liquid