Please login to your online account to display your discounted pricing

Melperone hydrochloride, Tocris

5-HT2A/D2 receptor antagonist; neuroleptic

$116.01 - $460.42

Chemical Identifiers

Chemical Name or Material Melperone hydrochloride
Assay Percent Range >99%
Molecular Formula C16H22FNO·HCl
Formula Weight 299.81
Synonym 1-(4-Fluorophenyl)-4-(4-methyl-1-piperidinyl)-1-butanone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

249550

 
r & d systems
RDS249550
50mg Each for $460.42

249510

 
r & d systems
RDS249510
10mg Each for $116.01
Description & Specifications

Specifications

Formula Weight 299.81
Quantity 50mg
Chemical Name or Material Melperone hydrochloride
Assay Percent Range >99%
Molecular Formula C16H22FNO·HCl
CAS 1622-79-3
Synonym 1-(4-Fluorophenyl)-4-(4-methyl-1-piperidinyl)-1-butanone hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature