missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ MELK Recombinant Human Protein
MELK is a serine/threonione kinase which is related to the AMPK family.
$2645.00 - $11260.00
Specifications
Accession Number | NP_055606.1 |
---|---|
Formulation | 50mM Tris, pH 7.5, 50% glycerol, 0.5mM EDTA, 0.02% Triton X-100, 150mM NaCl, 2mM DTT |
Molecular Weight (g/mol) | 66.8kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR8926B
|
Invitrogen™
PR8926B |
100μg |
Each for $2,645.00
|
|
|||||
PV4824
|
Invitrogen™
PV4824 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
MELK has been linked to the cell cycle. It has been shown to be overexpressed in cancer and has been identified has a potential target for cancer therapeutics.
Specifications
NP_055606.1 | |
66.8kDa | |
RUO - research use only | |
MELK | |
Human | |
GST Tag | |
Frozen |
50mM Tris, pH 7.5, 50% glycerol, 0.5mM EDTA, 0.02% Triton X-100, 150mM NaCl, 2mM DTT | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |