missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Melissic Acid, TCI America™

$106.95 - $551.55

Chemical Identifiers

CAS 506-50-3
Molecular Formula C30H60O2
Molecular Weight (g/mol) 452.808
MDL Number MFCD00002813
InChI Key VHOCUJPBKOZGJD-UHFFFAOYSA-N
Synonym melissic acid, 1-triacontanoic acid, n-triacontanoic acid, myricic acid, triacontoic acid, melissic acid a, unii-hlt2otq105, melissic acid,synthetic, hlt2otq105, ch3-ch2 28-cooh
PubChem CID 10471
ChEBI CHEBI:31003
IUPAC Name triacontanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M0595100MG View Documents TCI America
M0595100MG
100mg Each for $106.95
 
M05951G View Documents TCI America
M05951G
1g Each for $551.55
 
Specifications

Chemical Identifiers

506-50-3
452.808
VHOCUJPBKOZGJD-UHFFFAOYSA-N
10471
triacontanoic acid
C30H60O2
MFCD00002813
melissic acid, 1-triacontanoic acid, n-triacontanoic acid, myricic acid, triacontoic acid, melissic acid a, unii-hlt2otq105, melissic acid,synthetic, hlt2otq105, ch3-ch2 28-cooh
CHEBI:31003
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

White-Yellow
100mg
C30H60O2
melissic acid, 1-triacontanoic acid, n-triacontanoic acid, myricic acid, triacontoic acid, melissic acid a, unii-hlt2otq105, melissic acid,synthetic, hlt2otq105, ch3-ch2 28-cooh
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
452.808
CHEBI:31003
Crystal-Powder at 20°C
Melissic Acid
93°C
506-50-3
MFCD00002813
VHOCUJPBKOZGJD-UHFFFAOYSA-N
triacontanoic acid
10471
452.81
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

TSCA : No