missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Melissic Acid | |
93°C | |
C30H60O2 | |
1 g | |
VHOCUJPBKOZGJD-UHFFFAOYSA-N | |
triacontanoic acid | |
10471 | |
452.81 | |
Crystalline Powder |
506-50-3 | |
White-Yellow | |
MFCD00002813 | |
melissic acid, 1-triacontanoic acid, n-triacontanoic acid, myricic acid, triacontoic acid, melissic acid a, unii-hlt2otq105, melissic acid,synthetic, hlt2otq105, ch3-ch2 28-cooh | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O | |
452.81 | |
CHEBI:31003 | |
≥95.0% (GC) |
Chemical Identifiers
506-50-3 | |
452.81 | |
VHOCUJPBKOZGJD-UHFFFAOYSA-N | |
10471 | |
triacontanoic acid |
C30H60O2 | |
MFCD00002813 | |
melissic acid, 1-triacontanoic acid, n-triacontanoic acid, myricic acid, triacontoic acid, melissic acid a, unii-hlt2otq105, melissic acid,synthetic, hlt2otq105, ch3-ch2 28-cooh | |
CHEBI:31003 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : No