Please login to your online account to display your discounted pricing

Alfa Aesar™ Melanotan II

Chemical Identifiers

Chemical Name or Material Melanotan II
CAS 121062-08-6
Formula Weight 1024.18
MDL Number MFCD06795842
Synonym (Acetyl-Nle4, Asp5, DPhe7, Lys10)-cyclo-a-MSH (4-10) amide; Ac-Nle-c[Asp-His-DPhe-Arg-Trp-Lys]-NH2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65115MCR

  SDS
alfa aesar™
J65115MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1024.18
Chemical Name or Material Melanotan II
Physical Form Solid
MDL Number MFCD06795842
CAS 121062-08-6
Synonym (Acetyl-Nle4, Asp5, DPhe7, Lys10)-cyclo-a-MSH (4-10) amide; Ac-Nle-c[Asp-His-DPhe-Arg-Trp-Lys]-NH2
Storage Note 1 -30° to -10°C
RTECS Number OL4225000
Color White to off-white
Quantity 1mg
Safety and Handling

-30° to -10°C