Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available wit the Thermo Scientific™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody.

$331.94 - $1,171.37

Specifications

Antigen Melanoma (gp100) Ab-1
Clone HMB45
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS364R7

 
thermo scientific™ lab vision™
MS364R7
7mL Each for $678.47

MS364S

 
thermo scientific™ lab vision™
MS364S
1mL Each for $1,171.37

MS364S0

 
thermo scientific™ lab vision™
MS364S0
100μL Each for $331.94

MS364S1

 
thermo scientific™ lab vision™
MS364S1
500μL Each for $766.96
Description & Specifications

Specifications

Antigen Melanoma (gp100) Ab-1
Clone HMB45
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Host Species Murine
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Extract of pigmented melanoma metastases from lymph nodes

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available wit the Thermo Scientific™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody.

Research Areas: Cancer Marker

Host Species: Mouse

Clone: HMB45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with dog and rat. Others not known.

Immunogen: Extract of pigmented melanoma metastases from lymph nodes

Molecular Weight: 100kDa

Positive Control: Melanoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

    Western Blotting