missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Melamine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC125350100
Specifications
Melamine | |
Authentic | |
1.573 | |
354°C | |
108-78-1 | |
C3H6N6 | |
26, 245 | |
cyanuramide, cyanuric triamide, cyanurotriamide, cyanurotriamine, hicophor pr, isomelamine, melamine, teoharn, theoharn, triaminotriazine | |
JDSHMPZPIAZGSV-UHFFFAOYSA-N | |
1,3,5-triazine-2,4,6-triamine | |
7955 | |
126.12 | |
99% |
1.5730g/mL | |
Plastic drum | |
White | |
10kg | |
99% | |
MFCD00006055 | |
15, 5880 | |
Solubility in water: 3g/L (20°C). Other solubilities: 0.6g/L in ethanol (20°C),sparingly soluble in glycol,glycerol,pyridine,insoluble in ether,benzene,carbon tetrachloride | |
C1(=NC(=NC(=N1)N)N)N | |
126.12 | |
CHEBI:27915 | |
Fine Crystalline Powder |
Chemical Identifiers
108-78-1 | |
126.12 | |
JDSHMPZPIAZGSV-UHFFFAOYSA-N | |
7955 | |
1,3,5-triazine-2,4,6-triamine |
C3H6N6 | |
MFCD00006055 | |
cyanuramide, cyanuric triamide, cyanurotriamide, cyanurotriamine, hicophor pr, isomelamine, melamine, teoharn, theoharn, triaminotriazine | |
CHEBI:27915 | |
C1(=NC(=NC(=N1)N)N)N |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber : 203-615-4