missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Melamine, 99%

Manufacturer:  Alfa Aesar™ A112950E

 View more versions of this product

Catalog No. AAA112950E


 

Description

Description

Applications
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.

Solubility
Slightly soluble in water, hot alcohol, glycerol, glycol and pyridine. Insoluble in ether, benzene and carbon tetrachloride.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Melamine
2500g
99%
MFCD00006055
14,5811
JDSHMPZPIAZGSV-UHFFFAOYSA-N
1,3,5-triazine-2,4,6-triamine
7955
126.12
~350°C (decomposition)
108-78-1
C3H6N6
124341
2,4,6-Triamino-1,3,5-triazine
C1(=NC(=NC(=N1)N)N)N
126.123
CHEBI:27915

Chemical Identifiers

108-78-1
126.123
JDSHMPZPIAZGSV-UHFFFAOYSA-N
7955
C3H6N6
MFCD00006055
2,4,6-Triamino-1,3,5-triazine
CHEBI:27915
Safety and Handling

Safety and Handling

RTECSNumber : OS0700000

TSCA : TSCA

SDS Documents