Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MEK-1 Ab-1, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in western blotting experiments with the Thermo Scientific™ MEK-1 Ab-1, Rabbit Polyclonal Antibody.

$268.08 - $1,285.06

Specifications

Antigen MEK-1 Ab-1
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1662P

 
thermo scientific™ lab vision™
RB1662P
Purified Ab with BSA and Azide 1mL Each for $1,192.98

RB1662P0

 
thermo scientific™ lab vision™
RB1662P0
Purified Ab with BSA and Azide 100μL Each for $268.08

RB1662P1

 
thermo scientific™ lab vision™
RB1662P1
Purified Ab with BSA and Azide 500μL Each for $720.82

RB1662P1ABX

 
thermo scientific™ lab vision™
RB1662P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $815.86

RB1662PABX

 
thermo scientific™ lab vision™
RB1662PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,285.06
Description & Specifications

Specifications

Antigen MEK-1 Ab-1
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Immunogen A synthetic peptide derived from the N-terminus of human Mek1 protein
Isotype IgG

Ensure accurate, reproducible results in western blotting experiments with the Thermo Scientific™ MEK-1 Ab-1, Rabbit Polyclonal Antibody.

Mek1/mitogen activated protein kinase kinase/ERK kinase 1/microtubule-associated protein 2 kinase is a dual specificity kinase and catalyses both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat, Mouse and Xenopus. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide derived from the N-terminus of human MEK-1 protein

Molecular Weight: 48kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO