Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MEK-1 Ab-1, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in western blotting experiments with the Thermo Scientific™ MEK-1 Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1662PABX

 View more versions of this product

Catalog No. RB1662PABX

  • / Each
 

Description & Specifications

Specifications

Antigen MEK-1 Ab-1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic peptide derived from the N-terminus of human Mek1 protein
Isotype IgG
Quantity 1mL
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human
Target Species Murine
Target Species Rat

Ensure accurate, reproducible results in western blotting experiments with the Thermo Scientific™ MEK-1 Ab-1, Rabbit Polyclonal Antibody.

Mek1/mitogen activated protein kinase kinase/ERK kinase 1/microtubule-associated protein 2 kinase is a dual specificity kinase and catalyses both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Rat, Mouse and Xenopus. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide derived from the N-terminus of human MEK-1 protein

Molecular Weight: 48kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO