Please login to your online account to display your discounted pricing

Integra™ Miltex™ MeisterHand™ Operating Scissors


For general utility applications

Manufacturer: integra™ miltex™  MLTMH56

 View more versions of this product

Catalog No. 12-460-670

  • / Each
 

Description & Specifications

Specifications

Blade Style Straight
Material Stainless Steel
Tip Style Sharp/Sharp
Grade Standard
Length (English) 5.5 in.
Length (Metric) 13.97cm
Type Surgical, precision scissors
For Use With (Application) Surgical, Precision