Please login to your online account to display your discounted pricing

Integra™ Miltex™ MeisterHand™ Operating Scissors


For general utility applications

Manufacturer: integra™ miltex™  MLTMH54

 View more versions of this product

Catalog No. 12-460-666

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Surgical, Precision
Blade Style Straight
Tip Style Sharp/Sharp
Type Surgical, precision scissors
Material Stainless Steel
Grade Standard
Length (English) 5 in.
Length (Metric) 12.7cm