Please login to your online account to display your discounted pricing

Integra™ Miltex™ MeisterHand™ McPherson Micro Iris Suturing Forceps


Tying platform measures 0.3 x 5mm

Manufacturer: integra™ miltex™  MLTMH18949

 View more versions of this product

Catalog No. 12-460-618

  • / Each
 

Description & Specifications

Specifications

Quantity (No. per Case) Each
Length (Metric) 8.89cm
Tip Style Straight
Length (English) 3.5 in.
Product Type MeisterHand McPherson Micro Iris Suturing Forceps
Finish Non-glare Matte