Integra™ Miltex™ MeisterHand™ Mayo Dissecting Scissors
distal cutting
Supplier:  Integra™ Miltex™ MH5120
 View more versions of this product
Catalog No. 12-460-637
Description
Description
  • Stainless steel
  • Non-glare matted finish
  • Sharp/sharp scissors available in straight and curved patterns
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue and lower blade has micro fine serrations to help prevent tissue slippage; black ring handles
Order Info
Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-sharp edge to nick or become irregular.
Specifications
Specifications
Dissecting Scissors
5.75 in.
Stainless Steel
Straight, Beveled
14.6 cm
Dissecting