Integra™ Miltex™ MeisterHand™ Iris Scissors
Offer outstanding distal cutting capability
Supplier: Integra™ Miltex™ MH5306
Description
- Sharp/sharp scissors available in straight and curved
- Tungsten carbids scissors have tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
- SuperCut scissors have a razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to help prevent tissue slippage
- Stainless steel
- Non-glaring matte finish
Order Info
Cold or chemical sterilization not recommended for Carb-N-Sert scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.
Specifications
Iris Scissors | |
Standard | |
11.43 cm | |
Surgical, ophthalmic |
Curved | |
4.5 in. | |
Stainless Steel |