missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Integra™ Miltex™ MeisterHand™ Hartman Mosquito Forceps

Pattern length 4 in. (10cm)

$100.69 - $100.69

Specifications

Length (English) 3.5 in.
Length (Metric) 8.89cm
Material Stainless Steel
Product Type MeisterHand Hartman Mosquito Forceps
Quantity (No. per Case) Each
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Tip Style Price Quantity  
Catalog Number Mfr. No. Tip Style Price Quantity  
12-460-708 Integra™ Miltex™
MLTMH724
Straight Each for $100.69
Specifications

Description

  • Stainless steel
  • Non-glare matte finish
  • Full serrated jaws

Description & Specifications

Specifications

3.5 in.
Stainless Steel
Each
8.89cm
MeisterHand Hartman Mosquito Forceps
Non-glare Matte