Please login to your online account to display your discounted pricing

Integra™ Miltex™ MeisterHand™ Foerster Sponge Forceps

Straight design

$131.03 - $131.03

Specifications

Length (English) 9.75 in.
Length (Metric) 24.8cm
Material Stainless Steel
Quantity (No. per Case) Each
Finish Non-glare Matte
View More Specs
Products
Catalog Number Mfr. No. Product Type Tip Style Price Quantity    

12-460-718

 
integra™ miltex™
MLTMH7600
MeisterHand Foerster Sponge Forceps Straight, Serrated Jaws Each for $131.03
Description & Specifications

Specifications

Length (English) 9.75 in.
Length (Metric) 24.8cm
Material Stainless Steel
Quantity (No. per Case) Each
Finish Non-glare Matte

  • Length: 9.75 in. (24.8cm)
  • Stainless steel
  • Non-glare matte finish