Please login to your online account to display your discounted pricing

Integra™ Miltex™ MeisterHand™ Dressing Forceps


Standard pattern serrated handles

Manufacturer: integra™ miltex™  MLTMH68

 View more versions of this product

Catalog No. 12-460-702

  • / Each
 

Description & Specifications

Specifications

Tip Style Serrated
Material Stainless Steel
Product Type MeisterHand Dressing Forceps
Length (English) 5.5 in.
Quantity (No. per Case) Each
Finish Non-glare Matte
Length (Metric) 13.97cm