Fisherbrand™ Thermal Pouches with Zipper Closure
Fisherbrand Thermal Pouches with Zipper Closure keep temperature sensitive pharmaceutical products like insulin, vaccines and specimens cold for extended periods of time without ice
$1,112.03 - $1,619.70
Specifications
Closure Type Clear Zipper
Color Metalized PET Silver Exterior
Material Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
Quantity 200
Type Thermal Mailer
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
07-100-108 View Documents Fisherbrand™
KG1210PL00UZ182H
12in X 10in Case of 200 Each for $1,559.27
07-100-109 View Documents Fisherbrand™
KG0710PL00UZLN2H
7 x 10 in. Case of 200 Each for $1,112.03
07-100-107 View Documents Fisherbrand™
KG1512PL00UZ122H
15 in. X 12 in. Case of 200 Each for $1,619.70
Description
Description
Features:
  • Outer reflective surface
  • Inner food grade PR film
  • Micro cell foam insulation
  • Metallized outer film reflects radiant heat
  • Feature a double track zipper for ease of use
  • Use with gel pack, ice or dry ice for longer cooling time
  • Made in USA
Order Info
Case quantity only 200 per case
Warranty
Warranty against defects in material or workmanship.
Specifications
Specifications
Clear Zipper
Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
Thermal Mailer
Metalized PET Silver Exterior
200