missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Medroxyprogesterone Acetate, TCI America™

$55.95 - $167.88

Chemical Identifiers

CAS 71-58-9
Molecular Formula C24H34O4
Molecular Weight (g/mol) 386.532
MDL Number MFCD00010483
InChI Key PSGAAPLEWMOORI-PEINSRQWSA-N
Synonym depo-provera
PubChem CID 6279
ChEBI CHEBI:6716
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19641G View Documents Tci America
M19641G
1g Each for $55.95
 
M19645G View Documents Tci America
M19645G
5g Each for $167.88
 
Specifications

Chemical Identifiers

71-58-9
386.532
PSGAAPLEWMOORI-PEINSRQWSA-N
6279
[(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
C24H34O4
MFCD00010483
depo-provera
CHEBI:6716
CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C

Specifications

White
1g
C24H34O4
depo-provera
CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C
386.532
CHEBI:6716
Crystal-Powder at 20°C
Medroxyprogesterone Acetate
206°C
71-58-9
MFCD00010483
PSGAAPLEWMOORI-PEINSRQWSA-N
[(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
6279
386.53
≥93.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1470

RTECSNumber : TU5010000

TSCA : Yes