Medium Point Forceps
Steel construction ensures years of service in every kind of dissecting work.
$6.10 - $6.10
Specifications
Length (English) 3.98 in.
Length (Metric) 10.1cm
Quantity (No. per Case) Each
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Style Material Product Type Tip Style Price Quantity  
Catalog Number Mfr. No. Style Material Product Type Tip Style Price Quantity  
S17325 Fisher Science Education
SF5
Straight Chrome Plated Steel Forceps Straight Medium Point Each for $6.10
Case of 12 Each for $72.90
S17323 Fisher Science Education
MFM1212C
Curved Stainless Steel Dissecting Forceps Curved Medium Point Each for $6.10
Case of 12 Each for $72.90
Specifications
Description
  • Highly polished, chrome-plated steel forceps have file-cut, medium points for holding larger sections
  • Length: 3.98 in. (101mm)
Description & Specifications
Specifications
3.98 in.
10.1cm
Each