missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Medegen Saf-T Tainer™ Biomedical Waste Container

A plastic liner sandwiched between corrugated walls makes this container leak and puncture resistant

Manufacturer:  Medegen 102005

 View more versions of this product

Catalog No. 22-278-116



Description

Description

  • Rectangular, easy-to-assemble container stacks for convenient storage until incinerated
  • Integral handles for safe handling
  • Unassembled containers store flat to conserve space

Specifications

Specifications

Infectious Waste with Biohazard Symbol
12 in.
Red, Black
30.48 cm
Plastic
12 in.
30.48 cm
5 gal.
Biomedical Waste Container
30.4 cm
30.48 in.
SDS
Documents
Product Certifications
Promotions

Promotions