Please login to your online account to display your discounted pricing

MEDICA 16, Tocris™

FFA1 (GPR40) agonist; Also inhibits ATP citrate lyase

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material MEDICA 16
Assay Percent Range >98%
Molecular Formula C20H38O4
Formula Weight 342.51
Synonym 3,3,14,14-Tetramethylhexadecanedioic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

461510

 
r & d systems
RDS461510
10mg Each for $201.36

461550

 
r & d systems
RDS461550
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 342.51
Quantity 10mg
CAS 87272-20-6
Synonym 3,3,14,14-Tetramethylhexadecanedioic acid
Molecular Formula C20H38O4
Assay Percent Range >98%
Chemical Name or Material MEDICA 16
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C