missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Meclofenamic acid sodium salt, 99+%

Inhibits 5-lipoxygenase and cyclooxygenase; NSAID

Manufacturer:  Alfa Aesar™ J6048403

 View more versions of this product

Catalog No. AAJ6048403



Specifications

Specifications

Meclofenamic acid sodium salt
1g
C14H13Cl2NNaO2
meclofenamate sodium
JKTYCTJLWZYQGM-UHFFFAOYSA-N
2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium
131673979
Crystalline Solid
White
6385-02-0
MFCD00077376
Soluble in water at 50mg/ml; Also soluble in DMF, DMSO or ethanol
[HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na]
321.153
318.13
≥99%

Chemical Identifiers

6385-02-0
321.153
JKTYCTJLWZYQGM-UHFFFAOYSA-N
131673979
[HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na]
C14H13Cl2NNaO2
MFCD00077376
meclofenamate sodium
2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium
Safety and Handling

Safety and Handling

EINECSNumber : 228-983-3

RTECSNumber : CB2975500

TSCA : No

Recommended Storage : Store at -20°C

Documents