Please login to your online account to display your discounted pricing

Alfa Aesar™ Mecillinam, 95%

Effective against gram negative bacteria

Chemical Identifiers

Chemical Name or Material Mecillinam
CAS 32887-01-7
Formula Weight 325.43
MDL Number MFCD00056869
Merck Index 14,389
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66774MC

  SDS
alfa aesar™
J66774MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 325.43
CAS 32887-01-7
Chemical Name or Material Mecillinam
Assay 95%
MDL Number MFCD00056869
Color White
Physical Form Powder
Merck Index 14,389
Solubility Information Soluble in water at approximately 1mg/mL
Boiling Point 551°C
Melting Point 156°C
Storage Note 1 2° to 8°C
Quantity 100mg
RTECS Number XI0185000
Safety and Handling

2° to 8°C