missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mecarbinate 98.0+%, TCI America™
$48.11 - $137.70
Chemical Identifiers
CAS | 15574-49-9 |
---|---|
Molecular Formula | C13H15NO3 |
Molecular Weight (g/mol) | 233.267 |
MDL Number | MFCD00451373 |
InChI Key | YTBNTDMBGXAOCG-UHFFFAOYSA-N |
Synonym | Ethyl 5-Hydroxy-1,2-dimethylindole-3-carboxylate, 5-Hydroxy-1,2-dimethylindole-3-carboxylic Acid Ethyl Ester |
PubChem CID | 616236 |
IUPAC Name | ethyl 5-hydroxy-1,2-dimethylindole-3-carboxylate |
SMILES | CCOC(=O)C1=C(N(C2=C1C=C(C=C2)O)C)C |
Chemical Identifiers
15574-49-9 | |
233.267 | |
YTBNTDMBGXAOCG-UHFFFAOYSA-N | |
616236 | |
CCOC(=O)C1=C(N(C2=C1C=C(C=C2)O)C)C |
C13H15NO3 | |
MFCD00451373 | |
Ethyl 5-Hydroxy-1,2-dimethylindole-3-carboxylate, 5-Hydroxy-1,2-dimethylindole-3-carboxylic Acid Ethyl Ester | |
ethyl 5-hydroxy-1,2-dimethylindole-3-carboxylate |
Specifications
15574-49-9 | |
White-Yellow | |
MFCD00451373 | |
Ethyl 5-Hydroxy-1,2-dimethylindole-3-carboxylate, 5-Hydroxy-1,2-dimethylindole-3-carboxylic Acid Ethyl Ester | |
CCOC(=O)C1=C(N(C2=C1C=C(C=C2)O)C)C | |
233.267 | |
233.27 | |
Crystalline Powder |
212°C | |
C13H15NO3 | |
1 g | |
YTBNTDMBGXAOCG-UHFFFAOYSA-N | |
ethyl 5-hydroxy-1,2-dimethylindole-3-carboxylate | |
616236 | |
≥98.0% (GC) | |
Mecarbinate |
Safety and Handling
RTECSNumber : NL6003600
TSCA : No