Mettler Toledo™ ME-TE Precision Balances
For routine weighing procedures with intuitive touchscreen operation to simplify tasks. Mettler Toledo ME-TE Balances offer essential functionality, solid performance, and include External Calibration.
Manufacturer:  Mettler Toledo™ 30243386
 View more versions of this product
Catalog No. 01-914-471
Description
Description
  • Intuitive touchscreen operation
  • Wide range of optional accessories to make weighing even easier
  • Contains FACT - Fully automatic temperature controlled internal adjustment
  • Built-in overload protection
  • Just plug in and start (printer, barcode reader
  • Built-in RS232, USB device
  • Bluetooth adapter available
  • 8 embedded applications, including statistics and formulation
  • ISO/GLP certified
  • Supports underfloor weighing
Specifications
Specifications
ME503TE Balance
ME503TE Balance
1.5 sec.
Touchscreen
1 mg
1mg
520g
520 g
Documents