Please login to your online account to display your discounted pricing

MDL 73005EF hydrochloride, Tocris

Potent and selective 5-HT1A partial agonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material MDL 73005EF hydrochloride
Assay Percent Range >98%
Molecular Formula C20H26N2O4·HCl
Formula Weight 394.9
Synonym 8-[2-(1, 4-Benzodioxan-2-ylmethylamino)ethyl]-8-azaspiro[4.5]decane-7, 9-dione hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

041150

 
r & d systems
041150
50mg Each for $762.58

041110

 
r & d systems
041110
10mg Each for $181.19
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 394.9
Chemical Name or Material MDL 73005EF hydrochloride
Assay Percent Range >98%
Synonym 8-[2-(1, 4-Benzodioxan-2-ylmethylamino)ethyl]-8-azaspiro[4.5]decane-7, 9-dione hydrochloride
Molecular Formula C20H26N2O4·HCl
CAS 102908-60-1
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature