Please login to your online account to display your discounted pricing

MDL 73005EF hydrochloride, Tocris


Potent and selective 5-HT1A partial agonist

Quantity 10mg

Manufacturer: r & d systems  041110

 View more versions of this product

Catalog No. 041110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 394.9
Chemical Name or Material MDL 73005EF hydrochloride
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
CAS 102908-60-1
Molecular Formula C20H26N2O4·HCl
Synonym 8-[2-(1, 4-Benzodioxan-2-ylmethylamino)ethyl]-8-azaspiro[4.5]decane-7, 9-dione hydrochloride

Specifications

Chemical Name or Material MDL 73005EF hydrochloride
Assay Percent Range >98%
Molecular Formula C20H26N2O4·HCl
Formula Weight 394.9
Synonym 8-[2-(1, 4-Benzodioxan-2-ylmethylamino)ethyl]-8-azaspiro[4.5]decane-7, 9-dione hydrochloride
Accessories

Store at Room Temperature