missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ MDL 72527

Polyamine oxidase (PAO) inhibitor

$199.90 - $838.64

Specifications

Product Type MDL 72527
Inhibitors MDL 72527
Molecular Weight 265.23
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
370910R Tocris™
RDS370910
10mg Each for $199.90
370950R Tocris™
RDS370950
50mg Each for $838.64
Specifications

Description

Polyamine oxidase (POA) inhibitor. Does not inhibit monoamine oxidase or D-Amino acid oxidase. Displays anticancer and neuroprotective activity in vivo.
  • Chemical Name: N1,N4-Di-2,3-butadienyl-1,4-butanediamine dihydrochloride
  • Formula: C12H20N2.2HCl
  • CAS Number: 93565-01-6
  • Solubility: Soluble to 100mM in water and to 75mM in DMSO
Description & Specifications

Specifications

MDL 72527
MDL 72527
265.23