Please login to your online account to display your discounted pricing

MDL 12330A hydrochloride, Tocris

Adenylyl cyclase inhibitor; also cyclic nucleotide PDE inhibitor

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material MDL 12330A hydrochloride
Assay Percent Range >98%
Molecular Formula C23H36N2·HCl
Formula Weight 377.01
Synonym (±)-N-[(1R™, 2R™)-2-Phenylcyclopentyl]-azacyclotridec-1-en-2-amine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

143650

 
r & d systems
RDS143650
50mg Each for $598.33

143610

 
r & d systems
RDS143610
10mg Each for $142.08
Description & Specifications

Specifications

Formula Weight 377.01
Quantity 50mg
Chemical Name or Material MDL 12330A hydrochloride
Molecular Formula C23H36N2·HCl
CAS 40297-09-4
Synonym (±)-N-[(1R™, 2R™)-2-Phenylcyclopentyl]-azacyclotridec-1-en-2-amine hydrochloride
Assay Percent Range >98%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C