Please login to your online account to display your discounted pricing

MDL 11,939, Tocris


5-HT2A antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS087010

 View more versions of this product

Catalog No. 087010

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 295.42
Chemical Name or Material MDL 11, 939
Assay Percent Range >99%
Molecular Formula C20H25NO
CAS 107703-78-6
Recommended Storage Store at Room Temperature
Synonym α-Phenyl-1-(2-phenylethyl)-4-piperidinemethanol

Specifications

Chemical Name or Material MDL 11, 939
Assay Percent Range >99%
Molecular Formula C20H25NO
Formula Weight 295.42
Synonym α-Phenyl-1-(2-phenylethyl)-4-piperidinemethanol
Accessories

Store at Room Temperature