MCR Safety Klondike™ Safety Glasses
Manufacturer:  MCR Safety KD110
 View more versions of this product
Catalog No. 29-635-4367
Description
Description
  • Full orbital seal with dual lens design
  • Matte blue or black frame finish
  • Soft secure TPR nose pad
  • Extendable spatula temples
Specifications
Specifications
Exceeds ANSI Z87.1 Standards
Nylon
Adjustable
Scratch-resistant
Matte Black
Polycarbonate
Clear
Safety
Documents